3-Chloro-6-[(4,6-dimethylpyrimidin-2-yl)thio]pyridazine 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR7908-1G
APOSOR7908-1GEA 210 EUR
APOSOR7908-1G
3-Chloro-6-[(4,6-dimethylpyrimidin-2-yl)thio]pyridazine 95+%
3-Chloro-6-[(4,6-dimethylpyrimidin-2-yl)thio]pyridazine
Formule: C₁₀H₉ClN₄S
Smeltpunt: 161…163 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00214432
CAS nummer: 393183-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR