4-Bromo-2-(methylsulphonyl)benzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR7961-10G OR7961-1G OR7961-5G
APOSOR7961-10GEA 484 EUR
APOSOR7961-10G APOSOR7961-1G APOSOR7961-5G
4-Bromo-2-(methylsulphonyl)benzaldehyde
4-Bromo-2-(methylsulphonyl)benzaldehyde
Formule: C₈H₇BrO₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04037947
CAS nummer: 849035-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR