Benzo[b]thiophene-2-carboxaldehyde 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR8022-500MG
APOSOR8022-500MGEA 14.8 EUR
APOSOR8022-500MG
Benzo[b]thiophene-2-carboxaldehyde 96%
Benzo[b]thiophene-2-carboxaldehyde
Formule: C₉H₆OS
Kookpunt: 90 °C/0,1mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01075041
CAS nummer: 3541-37-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR