4-[(Trimethylsilyl)ethynyl]benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR8339-1G OR8339-5G
APOSOR8339-1GEA 80.4 EUR
APOSOR8339-1G APOSOR8339-5G
4-[(Trimethylsilyl)ethynyl]benzoic acid
4-[(Trimethylsilyl)ethynyl]benzoic acid
Formule: C₁₂H₁₄O₂Si
Smeltpunt: 177…178 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06203324
CAS nummer: 16116-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR