N,N-Dimethyl-2-[2-(aminomethyl)phenoxy]ethylamine 90%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR8685-1G OR8685-250MG
APOSOR8685-1GEA 311 EUR
APOSOR8685-1G APOSOR8685-250MG
N,N-Dimethyl-2-[2-(aminomethyl)phenoxy]ethylamine 90%
N,N-Dimethyl-2-[2-(aminomethyl)phenoxy]ethylamine
Formule: C₁₁H₁₈N₂O
Kookpunt: 89…91 °C (0,2 mm Hg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08690290
CAS nummer: 91215-97-3
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR