2-(1-Pyrrolidinyl)aniline 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR8694-1G OR8694-250MG OR8694-5G
APOSOR8694-1GEA 95.1 EUR
APOSOR8694-1G APOSOR8694-250MG APOSOR8694-5G
2-(1-Pyrrolidinyl)aniline 97%
2-(1-Pyrrolidinyl)aniline
Formule: C₁₀H₁₄N₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03563756
CAS nummer: 21627-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR