2,6-Dimethylbenzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR8724-100G OR8724-25G OR8724-5G
APOSOR8724-100GEA 172 EUR
APOSOR8724-100G APOSOR8724-25G APOSOR8724-5G
2,6-Dimethylbenzaldehyde
2,6-Dimethylbenzaldehyde
Formule: C₉H₁₀O
Kookpunt: 230 °C
Smeltpunt: 27…30 °C
Vlampunt: 96 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00128003
CAS nummer: 1123-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR