Di-tert-butyl disulphide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR8920-100ML OR8920-500ML
APOSOR8920-100MLEA 125 EUR
APOSOR8920-100ML APOSOR8920-500ML
Di-tert-butyl disulphide
Di-tert-butyl disulphide
Formule: C₈H₁₈S₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008838
CAS nummer: 110-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR