α,α'-Dibromo-o-xylene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR8961-100G OR8961-25G
APOSOR8961-100GEA 103 EUR
APOSOR8961-100G APOSOR8961-25G
α,α'-Dibromo-o-xylene
α,α'-Dibromo-o-xylene
Formule: C₈H₈Br₂
Smeltpunt: 91…96 °C
Dichtheid: 1,96 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000175
CAS nummer: 91-13-4
EINECS: 202-042-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR