3-(Ethanesulphonyl)aniline 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR900357-1G OR900357-250MG
APOSOR900357-1GEA 584 EUR
APOSOR900357-1G APOSOR900357-250MG
3-(Ethanesulphonyl)aniline 96%
3-(Ethanesulphonyl)aniline
Formule: C₈H₁₁NO₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11189664
CAS nummer: 83164-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR