2-Bromo-4-hydroxypyridine 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR901771-10G OR901771-1G OR901771-5G
APOSOR901771-10GEA 256 EUR
APOSOR901771-10G APOSOR901771-1G APOSOR901771-5G
2-Bromo-4-hydroxypyridine 96%
2-Bromo-4-hydroxypyridine
Formule: C₅H₄BrNO
Smeltpunt: 171 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00234040
CAS nummer: 36953-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR