5,6-Dichloropyrimidin-4-ol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR902633-1G OR902633-250MG
APOSOR902633-1GEA 99.3 EUR
APOSOR902633-1G APOSOR902633-250MG
5,6-Dichloropyrimidin-4-ol 95%
5,6-Dichloropyrimidin-4-ol
Formule: C₄H₂Cl₂N₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD24393897
CAS nummer: 88982-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR