1-(Benzo[d]oxazol-2-yl)ethanone 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR903710-1G OR903710-5G
APOSOR903710-1GEA 184 EUR
APOSOR903710-1G APOSOR903710-5G
1-(Benzo[d]oxazol-2-yl)ethanone 97%
1-(Benzo[d]oxazol-2-yl)ethanone
Formule: C₉H₇NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00223823
CAS nummer: 122433-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR