Methyl-6-bromo-1H-indole-2-carboxylate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR904285-1G OR904285-25G OR904285-5G
APOSOR904285-1GEA 29.2 EUR
APOSOR904285-1G APOSOR904285-25G APOSOR904285-5G
Methyl-6-bromo-1H-indole-2-carboxylate 97%
Methyl-6-bromo-1H-indole-2-carboxylate
Formule: C₁₀H₈BrNO₂
Smeltpunt: 170…190 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00590200
CAS nummer: 372089-59-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR