Amyl chloroformate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR904794-1G
APOSOR904794-1GEA 22 EUR
APOSOR904794-1G
Amyl chloroformate 97%
Amyl chloroformate
Formule: C₆H₁₁ClO₂
Kookpunt: 161 °C
Dichtheid: 1,03 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00058933
CAS nummer: 638-41-5
UN: 3277
ADR: 6.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR