7-Chloro-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR905796-10G OR905796-1G OR905796-5G
APOSOR905796-10GEA 342 EUR
APOSOR905796-10G APOSOR905796-1G APOSOR905796-5G
7-Chloro-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine 95%
7-Chloro-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Formule: C₆H₅ClN₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00227619
CAS nummer: 24415-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR