4-tert-Butyl-2-nitrophenol 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR905846-100G OR905846-25G
APOSOR905846-100GEA 208 EUR
APOSOR905846-100G APOSOR905846-25G
4-tert-Butyl-2-nitrophenol 98%
4-tert-Butyl-2-nitrophenol
Formule: C₁₀H₁₃NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00191888
CAS nummer: 3279-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR