Hexyl isocyanide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR906151-1G
APOSOR906151-1GEA 1290 EUR
APOSOR906151-1G
Hexyl isocyanide 95%
Hexyl isocyanide
Formule: C₇H₁₃N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00065203
CAS nummer: 15586-23-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR