N-Allyl-2-pyrrolidinone 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR906382-1G OR906382-5G
APOSOR906382-1GEA 74.6 EUR
APOSOR906382-1G APOSOR906382-5G
N-Allyl-2-pyrrolidinone 95%
N-Allyl-2-pyrrolidinone
Formule: C₇H₁₁NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00152226
CAS nummer: 2687-97-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR