4-(Acetamidomethyl)benzoic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR907934-1G OR907934-5G
APOSOR907934-1GEA 271 EUR
APOSOR907934-1G APOSOR907934-5G
4-(Acetamidomethyl)benzoic acid 98%
4-(Acetamidomethyl)benzoic acid
Formule: C₁₀H₁₁NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06655692
CAS nummer: 1205-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR