3-Chloro-5-methylbenzaldehyde 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR907959-1G OR907959-5G
APOSOR907959-1GEA 32.2 EUR
APOSOR907959-1G APOSOR907959-5G
3-Chloro-5-methylbenzaldehyde 98%
3-Chloro-5-methylbenzaldehyde
Formule: C₈H₇ClO
MW: 154.594 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11553565
CAS nummer: 103426-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR