Benzyl 3-chloropropanoate 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR908220-1G OR908220-5G
APOSOR908220-1GEA 146 EUR
APOSOR908220-1G APOSOR908220-5G
Benzyl 3-chloropropanoate 96%
Benzyl 3-chloropropanoate
Formule: C₁₀H₁₁ClO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD20542317
CAS nummer: 6282-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR