5-Bromo-2-methylbenzaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR909370-5G
APOSOR909370-5GEA 92.1 EUR
APOSOR909370-5G
5-Bromo-2-methylbenzaldehyde 97%
5-Bromo-2-methylbenzaldehyde
Formule: C₈H₇BrO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05662390
CAS nummer: 90050-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR