3-(4-Chlorophenyl)isoxazole 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR909998-10G OR909998-1G OR909998-5G
APOSOR909998-10GEA 760 EUR
APOSOR909998-10G APOSOR909998-1G APOSOR909998-5G
3-(4-Chlorophenyl)isoxazole 95%
3-(4-Chlorophenyl)isoxazole
Formule: C₉H₆ClNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08446383
CAS nummer: 31301-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR