4-Aminobenzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR9100-25G OR9100-100G
APOSOR9100-25GEA 22 EUR
APOSOR9100-25G APOSOR9100-100G
4-Aminobenzonitrile
4-Aminobenzonitrile
Formule: C₇H₆N₂
Smeltpunt: 83…85 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007821
CAS nummer: 873-74-5
EINECS: 212-850-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR