2-N-Ethylamino-5-nitropyridine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR911051-1G OR911051-25G OR911051-5G
APOSOR911051-1GEA 146 EUR
APOSOR911051-1G APOSOR911051-25G APOSOR911051-5G
2-N-Ethylamino-5-nitropyridine 98%
2-N-Ethylamino-5-nitropyridine
Formule: C₇H₉N₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00023438
CAS nummer: 67730-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR