O-tert-Butyl-L-serine methyl ester hydrochloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR911982-100G OR911982-25G
APOSOR911982-100GEA 349 EUR
APOSOR911982-100G APOSOR911982-25G
O-tert-Butyl-L-serine methyl ester hydrochloride 97%
O-tert-Butyl-L-serine methyl ester hydrochloride
Formule: C₈H₁₇NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00077108
CAS nummer: 17114-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR