(S)-(+)-Methyl mandelate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: L(+)-Mandelic acid methyl ester, L(+)-2-Hydroxy-2-phenylacetic acid methyl ester, Methyl (S)-(+)-mandelate, L(+)-Methyl mandelate, (S)-(+)-Mandelic acid methyl ester, (S)-(+)-2-Hydroxy-2-phenylacetic acid methyl ester, L(+)-Methyl glycolate, (S)-(+)-Methyl glycolate

OR912288-100G
APOSOR912288-100GEA 113 EUR
APOSOR912288-100G
(S)-(+)-Methyl mandelate 98%
(S)-(+)-Methyl mandelate
Formule: C₉H₁₀O₃
MW: 166.18 g/mol
Smeltpunt: 58
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064246
CAS nummer: 21210-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR