(1-Methyl-5-phenyl-1H-pyrazol-3-yl)methylamine 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR9131-1G OR9131-250MG
APOSOR9131-1GEA 315 EUR
APOSOR9131-1G APOSOR9131-250MG
(1-Methyl-5-phenyl-1H-pyrazol-3-yl)methylamine 95+%
(1-Methyl-5-phenyl-1H-pyrazol-3-yl)methylamine
Formule: C₁₁H₁₃N₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08271937
CAS nummer: 869901-12-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR