7-Methyl-1H-indole-3-carboxylic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR913136-5G
APOSOR913136-5GEA 55.5 EUR
APOSOR913136-5G
7-Methyl-1H-indole-3-carboxylic acid 98%
7-Methyl-1H-indole-3-carboxylic acid
Formule: C₁₀H₉NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06203634
CAS nummer: 30448-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR