Allyl-1-pyrrolidinecarbodithioate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR916805-1G OR916805-25G OR916805-5G
APOSOR916805-1GEA 125 EUR
APOSOR916805-1G APOSOR916805-25G APOSOR916805-5G
Allyl-1-pyrrolidinecarbodithioate 95%
Allyl-1-pyrrolidinecarbodithioate
Formule: C₈H₁₃NS₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00135118
CAS nummer: 701-13-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR