Octahydrocyclopenta[c]pyrrole 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR916903-10G OR916903-1G OR916903-25G
APOSOR916903-10GEA 351 EUR
APOSOR916903-10G APOSOR916903-1G APOSOR916903-25G
Octahydrocyclopenta[c]pyrrole 98%
Octahydrocyclopenta[c]pyrrole
Formule: C₇H₁₃N
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 5661-03-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR