2,2-Dimethylbut-3-ynoic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR917188-1G
APOSOR917188-1GEA 175 EUR
APOSOR917188-1G
2,2-Dimethylbut-3-ynoic acid 95%
2,2-Dimethylbut-3-ynoic acid
Formule: C₆H₈O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03093337
CAS nummer: 56663-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR