5-Oxohexanoic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR917515-100G OR917515-25G
APOSOR917515-100GEA 267 EUR
APOSOR917515-100G APOSOR917515-25G
5-Oxohexanoic acid 98%
5-Oxohexanoic acid
Formule: C₆H₁₀O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004412
CAS nummer: 3128-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR