4-(1-Hydroxy-1-methylethyl)benzoic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR917681-1G OR917681-250MG
APOSOR917681-1GEA 716 EUR
APOSOR917681-1G APOSOR917681-250MG
4-(1-Hydroxy-1-methylethyl)benzoic acid 97%
4-(1-Hydroxy-1-methylethyl)benzoic acid
Formule: C₁₀H₁₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02185844
CAS nummer: 3609-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR