7-Chloro-L-tryptophan 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR917825-250MG OR917825-500MG
APOSOR917825-250MGEA 593 EUR
APOSOR917825-250MG APOSOR917825-500MG
7-Chloro-L-tryptophan 97%
7-Chloro-L-tryptophan
Formule: C₁₁H₁₁ClN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09264322
CAS nummer: 73945-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR