4-(Bromomethyl)benzaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR9193-1G OR9193-250MG OR9193-5G
APOSOR9193-1GEA 24.9 EUR
APOSOR9193-1G APOSOR9193-250MG APOSOR9193-5G
4-(Bromomethyl)benzaldehyde 97%
4-(Bromomethyl)benzaldehyde
Formule: C₈H₇BrO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08271963
CAS nummer: 51359-78-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR