1-Phenyl-2-butanone 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR919635-5G
APOSOR919635-5GEA 191 EUR
APOSOR919635-5G
1-Phenyl-2-butanone 95%
1-Phenyl-2-butanone
Formule: C₆H₅C(O)CH₂CH₂CH₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009315
CAS nummer: 1007-32-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR