Methyl succinamate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methyl 4-amino-4-oxobutanoate, Succinamic Acid Methyl Ester

OR919943-5G
APOSOR919943-5GEA 48.3 EUR
APOSOR919943-5G
Methyl succinamate 95%
Methyl succinamate
Formule: C₅H₉NO₃
Kookpunt: 297,6 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08276419
CAS nummer: 53171-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR