Allyl isonicotinate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR920386-1G OR920386-25G OR920386-5G
APOSOR920386-1GEA 74.6 EUR
APOSOR920386-1G APOSOR920386-25G APOSOR920386-5G
Allyl isonicotinate 95%
Allyl isonicotinate
Formule: C₉H₉NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00059786
CAS nummer: 25635-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR