Butyl carbamate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR920407-100G OR920407-500G
APOSOR920407-100GEA 83.3 EUR
APOSOR920407-100G APOSOR920407-500G
Butyl carbamate 95%
Butyl carbamate
Formule: C₅H₁₁NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007967
CAS nummer: 592-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR