Ethanolamine 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR921532-100G OR921532-1KG
APOSOR921532-100GEA 22 EUR
APOSOR921532-100G APOSOR921532-1KG
Ethanolamine 99%
Ethanolamine
Formule: NH₂CH₂CH₂OH
Kookpunt: 170 °C
Smeltpunt: 10…11 °C
Dichtheid: 1,012
Vlampunt: 91 °C
MDL Nummer: MFCD00008183
CAS nummer: 141-43-5
UN: 2491
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR