1-[3-(Dimethylamino)propyl]-3-ethylurea 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR922815-5G
APOSOR922815-5GEA 199 EUR
APOSOR922815-5G
1-[3-(Dimethylamino)propyl]-3-ethylurea 98%
1-[3-(Dimethylamino)propyl]-3-ethylurea
Formule: C₈H₁₉N₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00027752
CAS nummer: 32897-26-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR