Allyl ethyl carbonate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR923998-5G
APOSOR923998-5GEA 104 EUR
APOSOR923998-5G
Allyl ethyl carbonate 95%
Allyl ethyl carbonate
Formule: C₆H₁₀O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049113
CAS nummer: 1469-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR