1-Amino-4-bromo-1H-pyrrole-2-carbonitrile 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR924899-1G OR924899-250MG
APOSOR924899-1GEA 833 EUR
APOSOR924899-1G APOSOR924899-250MG
1-Amino-4-bromo-1H-pyrrole-2-carbonitrile 95%
1-Amino-4-bromo-1H-pyrrole-2-carbonitrile
Formule: C₅H₄BrN₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD23105808
CAS nummer: 937161-86-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR