trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR925263-10G OR925263-1G OR925263-5G
APOSOR925263-10GEA 0 EUR
APOSOR925263-10G APOSOR925263-1G APOSOR925263-5G
trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 95%
trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid)
Formule: C₆H₆O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002702
CAS nummer: 3588-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR