Trichloroacetyl isocyanate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR925510-25G OR925510-5G
APOSOR925510-25GEA 431 EUR
APOSOR925510-25G APOSOR925510-5G
Trichloroacetyl isocyanate 95%
Trichloroacetyl isocyanate

  • May form small amount of sediment.

Formule: C₃Cl₃NO₂
Kookpunt: 80…85 °C
Dichtheid: 1,59 g/cm³ (25 °C)
Vlampunt: 65 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002033
CAS nummer: 3019-71-4
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR