4-Amino-3-methyl-pyrazole-1-carboxylic acid tert-butyl ester 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR925546-1G OR925546-5G
APOSOR925546-1GEA 220 EUR
APOSOR925546-1G APOSOR925546-5G
4-Amino-3-methyl-pyrazole-1-carboxylic acid tert-butyl ester 95%
4-Amino-3-methyl-pyrazole-1-carboxylic acid tert-butyl ester
Formule: C₉H₁₅N₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD23115361
CAS nummer: 847139-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR