5-Ethyl-2-hydroxybenzoic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR926276-100MG OR926276-1G OR926276-250MG
APOSOR926276-100MGEA 162 EUR
APOSOR926276-100MG APOSOR926276-1G APOSOR926276-250MG
5-Ethyl-2-hydroxybenzoic acid 95%
5-Ethyl-2-hydroxybenzoic acid
Formule: C₉H₁₀O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11203618
CAS nummer: 51-27-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR