4-Chloro-2-(methylamino)benzoic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR926983-1G OR926983-250MG
APOSOR926983-1GEA 438 EUR
APOSOR926983-1G APOSOR926983-250MG
4-Chloro-2-(methylamino)benzoic acid 95%
4-Chloro-2-(methylamino)benzoic acid
Formule: C₈H₈ClNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD17290555
CAS nummer: 54675-18-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR